1-(4-Chlorophenyl)-2-methylsulfonylethanone

Supplier: Matrix Scientific

Synonyms: 4'-Chloro-2-(methylsulphonyl)acetophenone

006898-1G 006898-5G
101791-488EA 67.69 USD
101791-488 101791-490
1-(4-Chlorophenyl)-2-methylsulfonylethanone
1-(4-Chlorophenyl)-2-methylsulfonylethanone

Hazard: Irritant

Formula: C₉H₉ClO₃S
MW: 232.69 g/mol
Melting Pt: 142…144 °C
MDL Number: MFCD00202209
CAS Number: 24437-48-7

Order Now

Specification Test Results

Melting point 142-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR