1-(3,5-Dichlorophenyl)-1,2-propandione

Supplier: Matrix Scientific

006897-1G 006897-5G
101791-484EA 63.08 USD
101791-484 101791-486
1-(3,5-Dichlorophenyl)-1,2-propandione
1-(3,5-Dichlorophenyl)-1,2-propandione

Hazard: Irritant

Formula: C₉H₆Cl₂O₂
Melting Pt: 45…46 °C
MDL Number: MFCD01941316
CAS Number: 386715-49-7

Order Now

Specification Test Results

Melting point 45-46°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR