1-(4-Nitrophenyl)-1,2-propandione

Supplier: Matrix Scientific

006896-1G 006896-5G
101791-480EA 63.08 USD
101791-480 101791-482
1-(4-Nitrophenyl)-1,2-propandione
1-(4-Nitrophenyl)-1,2-propandione

Hazard: Irritant

Formula: C₉H₇NO₄
MW: 193.16 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD01882876
CAS Number: 6159-25-7

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR