4-(Methylsulfonyl)phenylacetic acid

Supplier: Matrix Scientific

Synonyms: 2-(4-(Methylsulphonyl)phenyl)acetic acid

006879-1G 006879-5G
101791-436EA 29.23 USD
101791-436 101791-438
4-(Methylsulfonyl)phenylacetic acid
4-(Methylsulfonyl)phenylacetic acid

Hazard: Irritant

Formula: C₉H₁₀O₄S
Melting Pt: 137…138 °C
MDL Number: MFCD00216495
CAS Number: 90536-66-6

Order Now

Specification Test Results

Melting point 137-138°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR