2-Chloro-4-(methylsulfonyl)benzoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-methylsulfonylbenzoic acid, 2-Chloro-4-methylsulphonylbenzoic acid

006878-25G 006878-100G
101791-432EA 32.31 USD
101791-432 101791-434
2-Chloro-4-(methylsulfonyl)benzoic acid ≥97%
2-Chloro-4-(methylsulphonyl)benzoic acid

Hazard: Irritant

Formula: C₈H₇ClO₄S
MW: 234.66 g/mol
Melting Pt: 192-193°C
MDL Number: MFCD00216496
CAS Number: 53250-83-2

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR