2-(Methylsulfonyl)benzoic acid
Supplier: Matrix Scientific
006876-1G
006876-5G
101791-424EA
32.31
USD
101791-424
101791-426
2-(Methylsulfonyl)benzoic acid
2-(Methylsulphonyl)benzoic acid
Hazard: Irritant
Formula:
C₈H₈O₄S MW: 200.21 g/mol Melting Pt: 137-140°C |
MDL Number:
MFCD00216492 CAS Number: 33963-55-2 |
Specification Test Results
Melting point | 137-140°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...