2-Chloro-6-(trifluoromethyl)nicotinic acid

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid

006875-1G 006875-5G
101791-420EA 50.77 USD
101791-420 101791-422
2-Chloro-6-(trifluoromethyl)nicotinic acid
2-Chloro-6-(trifluoromethyl)nicotinic acid

Hazard: Irritant

Formula: C₇H₃ClF₃NO₂
Melting Pt: 120-122°C
MDL Number: MFCD01862658
CAS Number: 280566-45-2

Order Now

Specification Test Results

Melting point 120-122°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR