Methyl-3-formylindole-6-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 3-Formylindole-6-carboxylate

006870-1G 006870-5G
101791-402EA 36.92 USD
101791-402 101791-404
Methyl-3-formylindole-6-carboxylate
Methyl-3-formylindole-6-carboxylate

Hazard: Irritant

Formula: C₁₁H₉NO₃
MW: 203.197 g/mol
Melting Pt: 220…224 °C
MDL Number: MFCD00216479
CAS Number: 133831-28-4

Order Now

Specification Test Results

Melting point 220-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR