1-Hydroxy-6-methylsulfonylindole

Supplier: Matrix Scientific

006867-1G 006867-5G
101791-392EA 61.54 USD
101791-392 101791-394
1-Hydroxy-6-methylsulfonylindole
1-Hydroxy-6-methylsulfonylindole

Hazard: Irritant

Formula: C₉H₉NO₃S
MW: 211.24 g/mol
Melting Pt: 174…176 °C
MDL Number: MFCD00216482
CAS Number: 170492-47-4

Order Now

Specification Test Results

Melting point 174-176°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR