Ethyl-5-nitroindole-2-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-nitro-1H-indole-2-carboxylate

006866-1G 006866-5G
101791-388EA 21.54 USD
101791-388 101791-390
Ethyl-5-nitroindole-2-carboxylate ≥98%
Ethyl-5-nitroindole-2-carboxylate

Hazard: Irritant/Store in Freezer

Formula: C₁₁H₁₀N₂O₄
MW: 234.211 g/mol
Melting Pt: 223…225 °C
MDL Number: MFCD00216477
CAS Number: 16732-57-3

Order Now

Specification Test Results

Melting point 223-225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR