3-Formyl-2-methyl-5-nitroindole

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-nitro-1H-indole-3-carboxaldehyde

006864-1G 006864-5G
101791-380EA 63.08 USD
101791-380 101791-382
3-Formyl-2-methyl-5-nitroindole
3-Formyl-2-methyl-5-nitroindole

Hazard: Irritant

Formula: C₁₀H₈N₂O₃
MW: 204.19 g/mol
Melting Pt: 305…307 °C (dec.)
MDL Number: MFCD00216481
CAS Number: 3558-17-6

Order Now

Specification Test Results

Melting point 305-307°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR