2-(4-Chloro-2-nitrophenyl)malondialdehyde
Supplier: Matrix Scientific
006863-1G
006863-5G
101791-376EA
96.92
USD
101791-376
101791-378
2-(4-Chloro-2-nitrophenyl)malondialdehyde
2-(4-Chloro-2-nitrophenyl)malondialdehyde
Hazard: Irritant
Formula:
C₉H₆ClNO₄ Melting Pt: 140…143 °C |
MDL Number:
MFCD00216498 CAS Number: 205744-82-7 |
Specification Test Results
Melting point | 140-143°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...