2-(4-Pyridyl)malondialdehyde

Supplier: Matrix Scientific

006861-1G 006861-5G
101791-368EA 63.08 USD
101791-368 101791-370
2-(4-Pyridyl)malondialdehyde
2-(4-Pyridyl)malondialdehyde

Hazard: Irritant

Formula: C₈H₇NO₂
Melting Pt: >320 °C
MDL Number: MFCD00216535
CAS Number: 51076-46-1

Order Now

Specification Test Results

Melting point >320°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR