2-(2-Pyridyl)malondialdehyde

Supplier: Matrix Scientific

006860-1G 006860-5G
101791-364EA 60 USD
101791-364 101791-366
2-(2-Pyridyl)malondialdehyde
2-(2-Pyridyl)malondialdehyde

Hazard: Irritant

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Melting Pt: 202…205 °C
MDL Number: MFCD00216529
CAS Number: 212755-83-4

Order Now

Specification Test Results

Melting point 202-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR