2-(2-Quinolyl)malondialdehyde

Supplier: Matrix Scientific

006859-1G 006859-5G
101791-360EA 67.69 USD
101791-360 101791-362
2-(2-Quinolyl)malondialdehyde
2-(2-Quinolyl)malondialdehyde

Hazard: Irritant

Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Melting Pt: 200…202 °C (dec.)
MDL Number: MFCD01569245
CAS Number: 40070-84-6

Order Now

Specification Test Results

Melting point 200-202°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR