2-(4-Pyrimidyl)malondialdehyde
Supplier: Matrix Scientific
006858-1G
006858-5G
101791-356EA
381.54
USD
101791-356
101791-358
2-(4-Pyrimidyl)malondialdehyde
2-(4-Pyrimidyl)malondialdehyde
Hazard: Irritant
Formula:
C₇H₆N₂O₂ Melting Pt: 230 °C (dec.) |
CAS Number:
28648-78-4 |
Specification Test Results
Melting point | 230°C(dec) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...