2-(2-Benzoxazolyl)malondialdehyde

Supplier: Matrix Scientific

006857-1G 006857-5G
101791-352EA 72.31 USD
101791-352 101791-354
2-(2-Benzoxazolyl)malondialdehyde
2-(2-Benzoxazolyl)malondialdehyde

Hazard: Irritant

Formula: C₁₀H₇NO₃
Melting Pt: 230…232 °C (dec.)
MDL Number: MFCD00216526
CAS Number: 40071-07-6

Order Now

Specification Test Results

Melting point 230-232°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR