2-(2-Quinoxalinyl)malondialdehyde

Supplier: Matrix Scientific

006856-1G 006856-5G
101791-348EA 66.15 USD
101791-348 101791-350
2-(2-Quinoxalinyl)malondialdehyde
2-(2-Quinoxalinyl)malondialdehyde

Hazard: Irritant

Formula: C₁₁H₈N₂O₂
Melting Pt: 197…199 °C
MDL Number: MFCD00216534
CAS Number: 205744-84-9

Order Now

Specification Test Results

Melting point 197-199°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR