2-(4-Quinolyl)malondialdehyde sesquihydrate
Supplier: Matrix Scientific
006855-1G
006855-5G
101791-344EA
86.15
USD
101791-344
101791-346
2-(4-Quinolyl)malondialdehyde sesquihydrate
2-(4-Quinolyl)malondialdehyde sesquihydrate
Hazard: Irritant/Stored under Argon
Formula:
C₂₄H₂₄N₂O₇ Melting Pt: >150 °C (dec.) |
MDL Number:
MFCD09064915 CAS Number: 40070-86-8 |
Specification Test Results
Melting point | >150°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...