Ethyl-7-nitroindole-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 7-nitro-1H-indole-2-carboxylate

006854-1G 006854-5G
101791-340EA 67.69 USD
101791-340 101791-342
Ethyl-7-nitroindole-2-carboxylate
Ethyl-7-nitroindole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₄
MW: 234.211 g/mol
Melting Pt: 90…92 °C
MDL Number: MFCD00216475
CAS Number: 6960-46-9

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR