2-(4-Methoxyphenyl)malondialdehyde

Supplier: Matrix Scientific

006850-1G 006850-5G
101791-322EA 53.85 USD
101791-322 101791-324
2-(4-Methoxyphenyl)malondialdehyde
2-(4-Methoxyphenyl)malondialdehyde

Hazard: Irritant

Formula: C₁₀H₁₀O₃
Melting Pt: 145…148 °C
MDL Number: MFCD00216504
CAS Number: 65192-28-1

Order Now

Specification Test Results

Melting point 145-148°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR