2-(4-Methylsulfonyl-2-nitrophenyl)malondialdehyde

Supplier: Matrix Scientific

006849-1G 006849-5G
101791-318EA 49.23 USD
101791-318 101791-320
2-(4-Methylsulfonyl-2-nitrophenyl)malondialdehyde
2-(4-Methylsulfonyl-2-nitrophenyl)malondialdehyde

Hazard: Irritant

Formula: C₁₀H₉NO₆S
MW: 271.25 g/mol
Melting Pt: 215…217 °C (dec.)
MDL Number: MFCD00216509
CAS Number: 197251-71-1

Order Now

Specification Test Results

Melting point 215-217°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR