2-(2-Nitrophenyl)malondialdehyde

Supplier: Matrix Scientific

006847-1G 006847-5G
101791-310EA 53.85 USD
101791-310 101791-312
2-(2-Nitrophenyl)malondialdehyde
2-(2-Nitrophenyl)malondialdehyde

Hazard: Irritant

Formula: C₉H₇NO₄
MDL Number: MFCD00216503
CAS Number: 53868-44-3

Order Now

Specification Test Results

Melting point 119-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR