2-(5-Methoxy-2-nitrophenyl)malondialdehydemonohydrate

Supplier: Matrix Scientific

006846-1G 006846-5G
101791-306EA 43.08 USD
101791-306 101791-308
2-(5-Methoxy-2-nitrophenyl)malondialdehydemonohydrate
2-(5-Methoxy-2-nitrophenyl)malondialdehydemonohydrate

Hazard: Irritant

Formula: C₁₀H₁₁NO₆
MW: 241.2 g/mol
Melting Pt: 105…108 °C (dec.)
MDL Number: MFCD00216510
CAS Number: 205692-61-1

Order Now

Specification Test Results

Melting point 105-108°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR