4-(Methylsulfonyl)benzonitrile

Supplier: Matrix Scientific

006842-1G 006842-5G
101791-292EA 15.38 USD
101791-292 101791-294
4-(Methylsulfonyl)benzonitrile
4-(Methylsulphonyl)benzonitrile

Hazard: Irritant

Formula: C₈H₇NO₂S
Melting Pt: 140-142°C
MDL Number: MFCD00216489
CAS Number: 22821-76-7

Order Now

Specification Test Results

Melting point 140-142°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR