4-Fluoro-3-nitrophenyl methyl sulfone ≥97%

Supplier: Matrix Scientific

006841-5G 006841-25G
101791-288EA 58.46 USD
101791-288 101791-290
4-Fluoro-3-nitrophenyl methyl sulfone ≥97%
4-Fluoro-3-nitrophenyl methyl sulfone

Hazard: Irritant

Formula: C₇H₆FNO₄S
Melting Pt: 156…157 °C
MDL Number: MFCD00025066
CAS Number: 453-72-5

Order Now

Specification Test Results

Melting point 156-157°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR