6-Chloro-3-methyluracil ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-3-methylpyrimidine-2,4(1H,3H)-dione

006835-5G 006835-25G 006835-100G
101791-270EA 18.46 USD
101791-270 101791-272 101791-274
6-Chloro-3-methyluracil ≥97%
6-Chloro-3-methyluracil

Hazard: Irritant

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
Melting Pt: 278…280 °C (dec.)
MDL Number: MFCD01074837
CAS Number: 4318-56-3

Order Now

Specification Test Results

Melting point 278-280°C(d


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR