Methyl-2-(trifluoromethyl)pyridine-5-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 6-(trifluoromethyl)nicotinate

006832-1G 006832-5G
101791-258EA 44.62 USD
101791-258 101791-260
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate ≥98%
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate

Hazard: Irritant

Formula: C₈H₆F₃NO₂
MW: 205.13 g/mol
Melting Pt: 61…62 °C
MDL Number: MFCD03001346
CAS Number: 221313-10-6

Order Now

Specification Test Results

Melting point 61-62°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR