6-(Trifluoromethyl)nicotinamide
Supplier: Matrix Scientific
006831-1G
006831-5G
101791-254EA
106.15
USD
101791-254
101791-256
6-(Trifluoromethyl)nicotinamide
6-(Trifluoromethyl)nicotinamide
Hazard: Irritant
Formula:
C₇H₅F₃N₂O MW: 190.1200 g/mol Melting Pt: 185-187°C |
MDL Number:
MFCD01862646 CAS Number: 386715-35-1 |
Specification Test Results
Melting point | 185-187°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...