6-(Trifluormethyl)nicotinonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Cyano-2-(trifluoromethyl)pyridine, 6-(Trifluoromethyl)nicotinonitrile

006829-1G 006829-5G
101791-246EA 21.54 USD
101791-246 101791-248
6-(Trifluormethyl)nicotinonitrile ≥97%
6-(Trifluormethyl)nicotinonitrile

Hazard: Irritant

Formula: C₇H₃F₃N₂
MW: 172.1100 g/mol
Melting Pt: 37-40°C
MDL Number: MFCD03001347
CAS Number: 216431-85-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 37-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR