6-(Trifluoromethyl)pyridine-3-amidine hydrochloride

Supplier: Matrix Scientific

Synonyms: 6-(Trifluoromethyl)nicotinimidamide hydrochloride, 6-(Trifluoromethyl)nicotinamidine hydrochloride

006826-1G 006826-5G
101791-238EA 164.62 USD
101791-238 101791-240
6-(Trifluoromethyl)pyridine-3-amidine hydrochloride
6-(Trifluoromethyl)pyridine-3-amidine hydrochloride

Hazard: Irritant

Formula: C₇H₇ClF₃N₃
MW: 225.6 g/mol
Melting Pt: 191…193 °C
MDL Number: MFCD01862652
CAS Number: 221313-11-7

Order Now

Specification Test Results

Melting point 191-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR