2-Methylsulfonyl-1-(3-trifluoromethylphenyl)ethanone

Supplier: Matrix Scientific

Synonyms: 3-(Methylsulphonylacetyl)benzotrifluoride

006821-1G 006821-5G
101791-222EA 47.69 USD
101791-222 101791-224
2-Methylsulfonyl-1-(3-trifluoromethylphenyl)ethanone
2-Methylsulfonyl-1-(3-trifluoromethylphenyl)ethanone

Hazard: Irritant

Formula: C₁₀H₉F₃O₃S
MW: 266.24 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD00665597
CAS Number: 128306-96-7

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR