2-Methylsulfonyl-1-(pyridin-4-yl)ethanone

Supplier: Matrix Scientific

Synonyms: 3-[(Methylsulphonyl)acetyl]pyridine

006817-1G 006817-5G
101791-206EA 63.08 USD
101791-206 101791-208
2-Methylsulfonyl-1-(pyridin-4-yl)ethanone
2-Methylsulfonyl-1-(pyridin-4-yl)ethanone

Hazard: Irritant

Formula: C₈H₉NO₃S
MW: 199.23 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD01941302
CAS Number: 27302-94-9

Order Now

Specification Test Results

Melting point 115-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR