4-[(4,6-Dimethylpyrimidin-2-yl)thio]phenol

Supplier: Matrix Scientific

006811-1G 006811-5G
101791-186EA 135.38 USD
101791-186 101791-188
4-[(4,6-Dimethylpyrimidin-2-yl)thio]phenol
4-[(4,6-Dimethylpyrimidin-2-yl)thio]phenol

Hazard: Irritant

Formula: C₁₂H₁₂N₂OS
Melting Pt: 192…193 °C
MDL Number: MFCD00203057
CAS Number: 107718-34-3

Order Now

Specification Test Results

Melting point 192-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR