(Imidazo[2,1-b]benzothiazol-2-yl)methanol

Supplier: Matrix Scientific

006810-1G 006810-5G
101791-182EA 247.69 USD
101791-182 101791-184
(Imidazo[2,1-b]benzothiazol-2-yl)methanol
(Imidazo[2,1-b]benzothiazol-2-yl)methanol

Hazard: Irritant

Formula: C₁₀H₈N₂OS
Melting Pt: 172…174 °C
MDL Number: MFCD00203010
CAS Number: 114095-02-2

Order Now

Specification Test Results

Melting point 172-174°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR