7-Hydroxy-4-(pyridin-2-yl)coumarin

Supplier: Matrix Scientific

006808-1G 006808-5G
101791-174EA 47.69 USD
101791-174 101791-176
7-Hydroxy-4-(pyridin-2-yl)coumarin
7-Hydroxy-4-(pyridin-2-yl)coumarin

Hazard: Irritant

Formula: C₁₄H₉NO₃
Melting Pt: >250 °C (dec.)
MDL Number: MFCD01941327
CAS Number: 386704-10-5

Order Now

Specification Test Results

Melting point >250°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR