7-Hydroxycoumarin-4-acetic acid
Supplier: Matrix Scientific
Synonyms:
2-(7-Hydroxy-2-oxo-2H-chromen-4-yl)acetic acid, Umbelliferone-4-acetic acid
006807-1G
006807-5G
101791-170EA
58.46
USD
101791-170
101791-172
7-Hydroxycoumarin-4-acetic acid
7-Hydroxycoumarin-4-acetic acid
Hazard: Irritant
Formula:
C₁₁H₈O₅ MW: 220.18 g/mol Melting Pt: >195 °C (dec.) |
MDL Number:
MFCD00037563 CAS Number: 6950-82-9 |
Specification Test Results
Melting point | >195°C(dec) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...