7-Hydroxy-4-(3-trifluoromethylphenyl)coumarin

Supplier: Matrix Scientific

006805-1G 006805-5G
101791-160EA 40 USD
101791-160 101791-162
7-Hydroxy-4-(3-trifluoromethylphenyl)coumarin
7-Hydroxy-4-(3-trifluoromethylphenyl)coumarin

Hazard: Irritant

Formula: C₁₆H₉F₃O₃
MW: 306.24 g/mol
Melting Pt: >200 °C (dec.)
MDL Number: MFCD01941326
CAS Number: 386704-09-2

Order Now

Specification Test Results

Melting point >200°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR