7-Hydroxy-4-(pyridin-3-yl)coumarin

Supplier: Matrix Scientific

006804-1G 006804-5G
101791-156EA 47.69 USD
101791-156 101791-158
7-Hydroxy-4-(pyridin-3-yl)coumarin
7-Hydroxy-4-(pyridin-3-yl)coumarin

Hazard: Irritant

Formula: C₁₄H₉NO₃
Melting Pt: >230 °C (dec.)
MDL Number: MFCD01941325
CAS Number: 109512-76-7

Order Now

Specification Test Results

Melting point >230°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR