7-Hydroxy-4-(pyridin-4-yl)coumarin monohydrate

Supplier: Matrix Scientific

006803-1G 006803-5G
101791-152EA 47.69 USD
101791-152 101791-154
7-Hydroxy-4-(pyridin-4-yl)coumarin monohydrate
7-Hydroxy-4-(pyridin-4-yl)coumarin monohydrate

Hazard: Irritant

Formula: C₁₄H₁₁NO₄
Melting Pt: >300 °C (dec.)
MDL Number: MFCD01941324
CAS Number: 386704-08-1

Order Now

Specification Test Results

Melting point >300°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR