2,6-Dichloro-4-methylsulfonylphenol

Supplier: Matrix Scientific

006799-1G 006799-5G
101791-140EA 75.38 USD
101791-140 101791-142
2,6-Dichloro-4-methylsulfonylphenol
2,6-Dichloro-4-methylsulfonylphenol

Hazard: Irritant

Formula: C₇H₆Cl₂O₃S
MW: 241.09 g/mol
Melting Pt: 223…224 °C
MDL Number: MFCD00216500
CAS Number: 20951-05-7

Order Now

Specification Test Results

Melting point 223-224°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR