4-(Methylsulfonyl)phenol ≥97%

Supplier: Matrix Scientific

006798-5G 006798-25G
101791-136EA 16.92 USD
101791-136 101791-138
4-(Methylsulfonyl)phenol ≥97%
4-(Methylsulphonyl)phenol

Hazard: Irritant

Formula: C₇H₈O₃S
MW: 172.20 g/mol
Melting Pt: 91-93°C
MDL Number: MFCD00047806
CAS Number: 14763-60-1

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR