4-Phenoxyphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid

006790-5G 006790-25G
101791-124EA 20 USD
101791-124 101791-126
4-Phenoxyphenylboronic acid ≥97%
4-Phenoxyphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
Melting Pt: 141-145°C
MDL Number: MFCD00093312
CAS Number: 51067-38-0

Order Now

Specification Test Results

Melting point 141-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR