Benzo[b]furan-2-boronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: Benzofuran-2-boronic acid, 2-Benzofurylboronic acid

006753-1G 006753-5G
101791-018EA 36.92 USD
101791-018 101791-020
Benzo[b]furan-2-boronic acid ≥98%
Benzo[b]furan-2-boronic acid

Hazard: Irritant/Moisture Sensitive/Refrigerate

Formula: C₈H₇BO₃
Melting Pt: ca 197°C (dec)
MDL Number: MFCD00236019
CAS Number: 98437-24-2

Order Now

Specification Test Results

Melting point ca 197°C(de


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR