Methyl-2-naphthoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-naphthoate
006735-1G
006735-5G
101790-990EA
20
USD
101790-990
101790-992
Methyl-2-naphthoate ≥97%
Methyl 2-naphthoate
Hazard: Irritant
Formula:
C₁₂H₁₀O₂ Melting Pt: 71-74°C |
MDL Number:
MFCD00017345 CAS Number: 2459-25-8 |
Specification Test Results
Melting point | 71-74°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...