1-Dimethylamino-2-nitroethylene ≥98%

Supplier: Matrix Scientific

006729-1G 006729-5G
101790-978EA 46.15 USD
101790-978 101790-980
1-Dimethylamino-2-nitroethylene ≥98%
1-Dimethylamino-2-nitroethylene

Hazard: Irritant/Refrigerate

Formula: C₄H₈N₂O₂
MW: 116.12 g/mol
Melting Pt: 103…105 °C
MDL Number: MFCD00051519
CAS Number: 1190-92-7

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR