3-Aminothiophene-2-carboxamide ≥97%

Supplier: Matrix Scientific

006719-250MG 006719-1G
101790-956EA 23.08 USD
101790-956 101790-958
3-Aminothiophene-2-carboxamide ≥97%
3-Aminothiophene-2-carboxamide

Hazard: Irritant

Formula: C₅H₆N₂OS
MW: 142.18 g/mol
Melting Pt: 120-122°C
MDL Number: MFCD00052593
CAS Number: 147123-47-5

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR