4-Methyl-β-methyl-β-nitrostyrene ≥98%

Supplier: Matrix Scientific

006705-5G 006705-25G
101790-944EA 155.38 USD
101790-944 101790-946
4-Methyl-β-methyl-β-nitrostyrene ≥98%
4-Methyl-beta-methyl-beta-nitrostyrene

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD00024817
CAS Number: 52287-56-6

Order Now

Specification Test Results

Melting point 51-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR