Ethyl-4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate
Supplier: Matrix Scientific
Synonyms:
Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate
006588-1G
006588-5G
101790-804EA
72.31
USD
101790-804
101790-806
Ethyl-4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate
Ethyl 4-hydroxy-6-(trifluoromethoxy)quinoline-3-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₀F₃NO₄ MW: 301.2200 g/mol Melting Pt: 297…305 °C |
MDL Number:
MFCD00219847 CAS Number: 175203-85-7 |
Specification Test Results
Melting point | 297-305°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...