4-(Benzyloxy)benzonitrile ≥99%

Supplier: Matrix Scientific

006549-5G 006549-25G
101790-714EA 118.46 USD
101790-714 101790-716
4-(Benzyloxy)benzonitrile ≥99%
4-(Benzyloxy)benzonitrile

Hazard: Irritant

Formula: C₁₄H₁₁NO
Melting Pt: 97…98 °C
MDL Number: MFCD00079701
CAS Number: 52805-36-4

Order Now

Specification Test Results

Melting point 97-98°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR